Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083911.1 | NP_001077380.1 | 2 | 245 | ||
NM_019192.2 | NP_062065.2 | 2 | 173 | |||
GenBank mRNA | BC059137.1 | - | 2 | 174 | ||
BC072539.1 | - | 2 | 162 | 7104195 | ||
CB699091.1 | - | 2 | 245 | |||
CB745388.1 | - | 2 | 397 | |||
D25221.1 | - | 2 | 154 | |||
DN933732.1 | - | 2 | 493 | |||
M63574.1 | - | 133 |