Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019201.3 | NP_062074.2 | 4 | 656 | ||
XM_006251225.2 | XP_006251287.1 | 3 | 491 | |||
XM_006251226.2 | XP_006251288.1 | 3 | 491 | |||
XM_006251227.2 | XP_006251289.1 | 3 | 490 | |||
XM_006251228.2 | XP_006251290.1 | 3 | 491 | |||
XM_006251229.2 | XP_006251291.1 | 4 | 682 | |||
GenBank mRNA | AF067795.2 | - | 4 | 661 | ||
BC078778.1 | - | 4 | 667 | 7108845 |