Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019210.1 | NP_062083.1 | 2 | 204 | ||
XM_006257333.2 | XP_006257395.1 | 3 | 1432 | |||
XM_006257336.2 | XP_006257398.1 | 2 | 1321 | |||
XM_006257338.2 | XP_006257400.1 | 2 | 1724 | |||
XM_006257339.2 | XP_006257401.1 | 3 | 1431 | |||
XM_008773404.1 | XP_008771626.1 | 3 | 1434 | |||
GenBank mRNA | U33314.1 | - | 2 | 204 |