Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019214.1 | NP_062087.1 | 20 | 2801 | ||
XM_008764576.1 | XP_008762798.1 | 21 | 4125 | |||
XM_008764577.1 | XP_008762799.1 | 19 | 2892 | |||
GenBank mRNA | AF167412.1 | - | 20 | 2801 |