Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012309.4 | NP_036441.2 | 15 | 2025 | ||
NM_133266.4 | NP_573573.2 | 1 | 222 | |||
NR_110766.1 | 1 | 222 | ||||
XM_005277932.3 | XP_005277989.1 | 6 | 1030 | |||
XM_017017387.1 | XP_016872876.1 | 16 | 2130 | |||
XM_017017388.1 | XP_016872877.1 | 16 | 2152 | |||
XM_017017389.1 | XP_016872878.1 | 16 | 2325 | |||
XM_017017390.1 | XP_016872879.1 | 3 | 3263 | |||
GenBank mRNA | AB208025.1 | - | 15 | 1991 | ||
AB208026.1 | - | 6 | 813 | |||
AB208027.1 | - | 1 | 200 | |||
AF141901.1 | - | 1 | 114 | |||
BC093885.1 | - | 1 | 222 | |||
BC112097.1 | - | 1 | 222 | |||
BC113876.1 | - | 1 | 145 | |||
BC114484.1 | - | 1 | 144 | |||
BC143389.1 | - | 1 | 222 | |||
DQ152234.1 | - | 14 | 1913 |