Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019292.4 | NP_062165.2 | 2 | 99 | ||
XM_006232116.1 | XP_006232178.1 | 3 | 424 | |||
GenBank mRNA | AB030829.1 | - | 2 | 99 | ||
AB030830.1 | - | 2 | 99 | |||
AF037072.1 | - | 2 | 122 | |||
BC061980.1 | - | 2 | 134 | 5597975 | ||
FQ210528.1 | - | 2 | 147 | |||
FQ211459.1 | - | 2 | 146 | |||
FQ214846.1 | - | 2 | 144 | |||
FQ214929.1 | - | 2 | 149 | |||
FQ216182.1 | - | 2 | 145 | |||
FQ223846.1 | - | 2 | 145 | |||
M22413.1 | - | 2 | 254 |