Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232116.1 | XP_006232178.1 | 8 | 1287 | ||
GenBank mRNA | AB030829.1 | - | 7 | 962 | ||
AF037072.1 | - | 7 | 985 | |||
BC061980.1 | - | 7 | 997 | 5597975 | ||
FQ210528.1 | - | 7 | 1011 | |||
FQ211459.1 | - | 7 | 1009 | |||
FQ214846.1 | - | 7 | 1008 | |||
FQ214929.1 | - | 7 | 1014 | |||
FQ215392.1 | - | 6 | 824 | |||
FQ216182.1 | - | 7 | 1009 | |||
FQ223846.1 | - | 7 | 1009 |