Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cltc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019299.1 | NP_062172.1 | 2 | 173 | ||
XM_006247107.2 | XP_006247169.1 | 2 | 482 | |||
XM_006247108.2 | XP_006247170.1 | 2 | 481 | |||
XM_006247109.2 | XP_006247171.1 | 2 | 481 | |||
GenBank mRNA | J03583.1 | - | 2 | 173 |