Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005265.1 | NP_001005265.1 | 2 | 408 | ||
NM_001005330.1 | NP_001005330.1 | 408 | ||||
NM_019301.2 | NP_062174.1 | 2 | 408 | |||
GenBank mRNA | BC061736.1 | - | 2 | 177 | 5599318 | |
CB707374.1 | - | 2 | 360 | |||
D42114.1 | - | 2 | 217 | |||
D42115.1 | - | 2 | 153 | |||
L36532.1 | - | 2 | 176 |