Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gja6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019308.1 | NP_062181.1 | 1 | 692 | ||
XM_006256894.2 | XP_006256956.1 | 2 | 1069 |