Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004105.2 | NP_001004105.1 | 2 | 241 | ||
NM_001004106.2 | NP_001004106.1 | 2 | 241 | |||
NM_002082.3 | NP_002073.2 | 2 | 241 | |||
XM_006714859.2 | XP_006714922.1 | 2 | 153 | |||
XM_011534537.2 | XP_011532839.1 | 2 | 106 | |||
GenBank mRNA | AF040751.1 | - | 2 | 84 | ||
AF040752.1 | - | 2 | 84 | |||
BC009277.2 | - | 2 | 181 | 4053197 | ||
BC017272.1 | - | 2 | 130 | 4640283 | ||
BI258860.1 | - | 2 | 242 | |||
BX382886.2 | - | 2 | 239 | |||
L16862.1 | - | 2 | 143 |
Gene Symbol: GRK6P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295462.1 | - | 1 | 145 |