Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019377.1 | NP_062250.1 | 6 | 1058 | ||
GenBank mRNA | BC076502.1 | - | 6 | 1052 | 7100393 | |
D17446.1 | - | 6 | 1053 | |||
S55223.1 | - | 6 | 1053 | |||
S83440.1 | - | 6 | 1058 |