Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020076.2 | NP_064461.1 | 2, 3 | 252 | ||
XM_006239684.2 | XP_006239746.1 | 2, 3 | 1157 | |||
GenBank mRNA | BC085739.1 | - | 2, 3 | 206 | 7108551 | |
D44494.1 | - | 2, 3 | 253 | |||
FQ219702.1 | - | 2, 3 | 266 |