Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020093.1 | NP_064478.1 | 3 | 380 | ||
GenBank mRNA | AB039878.1 | - | 3 | 380 | ||
AF168004.1 | - | 3 | 400 | |||
AF210434.1 | - | 3 | 372 | |||
AF257234.1 | - | 2 | 310 | |||
AF343575.1 | - | 1 | 146 | |||
AF381277.1 | - | 3 | 382 | |||
AF381278.1 | - | 2 | 310 | |||
AF381279.1 | - | 2 | 310 | |||
AF381280.1 | - | 3 | 466 | |||
AF381281.1 | - | 2 | 310 | |||
GU322017.1 | - | 4 | 479 | |||
GU322019.1 | - | 3 | 323 | |||
GU322363.1 | - | 3 | 323 | |||
GU322364.1 | - | 3 | 323 | |||
GU345835.1 | - | 3 | 323 | |||
GU345836.1 | - | 3 | 323 | |||
KC774169.1 | - | 2 | 166 | |||
KC774170.1 | - | 2 | 166 | |||
KC774172.1 | - | 3 | 320 | |||
KC774173.1 | - | 2 | 166 | |||
KC774175.1 | - | 2 | 166 | |||
KC774177.1 | - | 2 | 166 |