Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311255.1 | NP_001298184.1 | 2 | 272 | ||
NM_001311256.1 | NP_001298185.1 | 2 | 272 | |||
NM_012388.3 | NP_036520.1 | 2 | 438 | |||
NR_132350.1 | 2 | 272 | ||||
NR_132351.1 | 2 | 438 | ||||
NR_132352.1 | 2 | 438 | ||||
NR_132356.1 | 2 | 140 | ||||
NR_132357.1 | 2 | 140 | ||||
GenBank mRNA | AF080470.1 | - | 2 | 342 | ||
AK057545.1 | - | 2 | 182 | |||
AK128626.1 | - | 2 | 241 | |||
AK315581.1 | - | 2 | 156 | |||
AL596952.1 | - | 2 | 211 | |||
BC004819.1 | - | 2 | 159 | 3450397 | ||
DA247079.1 | - | 2 | 158 | |||
DA468279.1 | - | 2 | 438 | |||
DA523523.1 | - | 2 | 86 | |||
DA859617.1 | - | 2 | 87 | |||
DB289156.1 | - | 2 | 83 |