Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021583.3 | NP_067594.1 | 3 | 629 | ||
GenBank mRNA | AB041998.1 | - | 3 | 618 | ||
AB048730.1 | - | 3 | 631 | |||
BC088101.1 | - | 3 | 587 | |||
BC105858.1 | - | 3 | 615 | 7456259 |