Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012394.3 | NP_036526.2 | 1, 2 | 133 | ||
GenBank mRNA | AF117237.1 | - | 1, 2 | 89 | ||
AF165883.1 | - | 1, 2 | 106 | |||
AK024531.1 | - | 1, 2 | 113 | |||
AK315736.1 | - | 1, 2 | 107 | |||
BC012464.1 | - | 1, 2 | 102 | 4538003 | ||
BC047042.1 | - | 1, 2 | 99 | 5541226 | ||
BI597083.1 | - | 1, 2 | 166 |