Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dio1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021653.3 | NP_067685.5 | 4 | 1572 | ||
GenBank mRNA | BC083557.1 | - | 4 | 1567 | 7110283 | |
X57999.1 | - | 4 | 1572 |