Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270931.1 | NP_001257860.1 | 5 | 628 | ||
NM_001270933.1 | NP_001257862.1 | 5 | 628 | |||
NM_022180.2 | NP_071516.2 | 4 | 588 | |||
XM_006235515.2 | XP_006235577.1 | 4 | 583 | |||
XM_006235516.2 | XP_006235578.1 | 4 | 583 | |||
GenBank mRNA | CK483260.1 | - | 4 | 417 | ||
D10554.1 | - | 4 | 464 | |||
EF193390.1 | - | 4 | 409 | |||
EF193392.1 | - | 4 | 390 | |||
FJ608816.1 | - | 5 | 628 | |||
FJ608817.1 | - | 5 | 628 | |||
FJ608818.1 | - | 5 | 628 | |||
FM042560.1 | - | 4 | 593 | |||
X57133.1 | - | 4 | 479 |