Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022182.1 | NP_071518.1 | 1 | 274 | ||
XM_006234903.2 | XP_006234965.1 | 2 | 1594 | |||
GenBank mRNA | AF295300.1 | - | 1 | 242 | ||
X56551.1 | - | 1 | 274 |