Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271057.1 | NP_001257986.1 | 3, 4 | 184 | ||
NM_022516.5 | NP_071961.2 | 3, 4 | 184 | |||
GenBank mRNA | BC061858.1 | - | 3, 4 | 179 | 5598495 | |
CB772933.1 | - | 3, 4 | 194 | |||
FN803753.1 | - | 3, 4 | 154 | |||
X60789.1 | - | 3, 4 | 147 | |||
X74565.1 | - | 3, 4 | 165 |