Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022583.1 | NP_072105.1 | 1 | 202 | ||
GenBank mRNA | AF159506.1 | - | 1 | 202 |