Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022928.1 | NP_075217.1 | 6 | 665 | ||
XM_006251360.1 | XP_006251422.1 | 6 | 501 | |||
XM_008770353.1 | XP_008768575.1 | 6 | 501 | |||
XM_008770355.1 | XP_008768577.1 | 6 | 501 | |||
GenBank mRNA | X97568.1 | - | 6 | 665 |