Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024359.1 | NP_077335.1 | 2 | 151 | ||
XM_006240196.2 | XP_006240258.1 | 2 | 464 | |||
XM_006240197.2 | XP_006240259.1 | 2 | 461 | |||
XM_006240198.2 | XP_006240260.1 | 2 | 460 | |||
XM_006240199.2 | XP_006240261.1 | 2 | 163 | |||
XM_008764695.1 | XP_008762917.1 | 2 | 248 | |||
GenBank mRNA | AF057308.1 | - | 2 | 151 | ||
Y09507.1 | - | 2 | 248 |