Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arntl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024362.2 | NP_077338.2 | 7, 8 | 764 | ||
XM_006230029.2 | XP_006230091.1 | 5, 6 | 522 | |||
XM_006230030.2 | XP_006230092.1 | 5, 6 | 522 | |||
XM_008759716.1 | XP_008757938.1 | 5, 6 | 462 | |||
GenBank mRNA | AB012600.1 | - | 2, 3 | 196 | ||
AF015953.1 | - | 2, 3 | 186 | |||
AF317669.2 | - | 2, 3 | 122 | |||
BC099833.1 | - | 7, 8 | 764 | 7936079 |