Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077491.1 | NP_001070959.1 | 4, 5 | 555 | ||
NM_001077492.1 | NP_001070960.1 | 3, 4 | 487 | |||
NM_012427.4 | NP_036559.1 | 3, 4 | 690 | |||
XM_011526702.1 | XP_011525004.1 | 3, 4 | 429 | |||
XM_011526703.2 | XP_011525005.1 | 4, 5 | 509 | |||
GenBank mRNA | AF168768.2 | - | 3, 4 | 506 | ||
AF435980.1 | - | 3, 4 | 661 | |||
AF435981.1 | - | 3, 4 | 458 | |||
AK223421.1 | - | 3, 4 | 638 | |||
AY279380.1 | - | 3, 4 | 464 | |||
AY279381.1 | - | 4, 5 | 532 | |||
AY359010.1 | - | 3, 4 | 690 | |||
AY461805.1 | - | 2, 3 | 348 | |||
BC008036.2 | - | 3, 4 | 621 | 2989806 | ||
BT006867.1 | - | 2, 3 | 337 |