Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291932.1 | NP_001278861.1 | 3, 4 | 342 | ||
NM_012429.4 | NP_036561.1 | 4, 5 | 418 | |||
NM_033382.2 | NP_203740.1 | 4, 5 | 418 | |||
GenBank mRNA | AB033012.2 | - | 4, 5 | 317 | ||
AK223587.1 | - | 4, 5 | 378 | |||
AK296540.1 | - | 3, 4 | 232 | |||
AK314390.1 | - | 4, 5 | 351 | |||
AL096881.1 | - | 4, 5 | 415 | |||
AY927439.1 | - | 3, 4 | 147 | |||
BC058915.1 | - | 4, 5 | 322 | 5178251 | ||
CR456571.1 | - | 4, 5 | 315 | |||
DC375707.1 | - | 3, 4 | 342 |