Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031579.2 | NP_113767.1 | 6 | 1844 | ||
XM_006244670.2 | XP_006244732.1 | 6 | 1726 | |||
XM_006244671.2 | XP_006244733.1 | 6 | 1709 | |||
GenBank mRNA | BC081772.1 | - | 5 | 1474 | 7124031 | |
BC097307.1 | - | 6 | 1844 | 7446277 | ||
FQ220156.1 | - | 1 | 146 | |||
L27843.1 | - | 6 | 1673 |
Gene Symbol: LOC103693189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766779.1 | XP_008765001.1 | 5 | 1643 |