Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031584.2 | NP_113772.1 | 3 | 737 | ||
GenBank mRNA | D83044.1 | - | 3 | 740 | ||
X98334.1 | - | 3 | 686 | |||
Y13154.1 | - | 3 | 687 |