Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012437.5 | NP_036569.1 | 4 | 457 | ||
NR_052019.1 | 3 | 410 | ||||
NR_052020.1 | 3 | 396 | ||||
GenBank mRNA | AF086837.1 | - | 4 | 367 | ||
AK024555.1 | - | 4 | 434 | |||
BC000761.2 | - | 4 | 386 | 2821705 | ||
BC004494.2 | - | 4 | 387 | 3830132 | ||
BE278772.1 | - | 3 | 311 | |||
BI117540.1 | - | 458 | ||||
BM051828.1 | - | 4 | 443 | |||
BT006753.1 | - | 4 | 367 | |||
CD671119.1 | - | 3 | 345 |