Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031587.1 | NP_113775.1 | 1, 2 | 144 | ||
GenBank mRNA | AY327502.1 | - | 1, 2 | 161 | ||
X70223.1 | - | 1, 2 | 144 |