Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031587.1 | NP_113775.1 | 5 | 825 | ||
GenBank mRNA | X70223.1 | - | 5 | 825 |