Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031599.2 | NP_113787.1 | 3, 4 | 837 | ||
XM_006236623.1 | XP_006236685.1 | 3, 4 | 355 | |||
XM_006236624.2 | XP_006236686.1 | 3, 4 | 875 | |||
XM_008762977.1 | XP_008761199.1 | 2, 3 | 469 | |||
GenBank mRNA | AF096835.1 | - | 3, 4 | 838 |