Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031600.1 | NP_113788.1 | 2, 3 | 278 | ||
GenBank mRNA | U73458.1 | - | 2, 3 | 278 | ||
Z50735.1 | - | 2, 3 | 252 |