Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031603.1 | NP_113791.1 | 2 | 200 | ||
GenBank mRNA | BC063163.1 | - | 2 | 178 | 6921685 | |
D30739.1 | - | 2 | 184 | |||
FQ220393.1 | - | 2 | 206 | |||
M84416.1 | - | 2 | 180 | |||
U53882.1 | - | 2 | 200 |