Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031609.1 | NP_113797.1 | 4 | 1048 | ||
XM_008764284.1 | XP_008762506.1 | 4 | 1012 | |||
GenBank mRNA | BC061833.1 | - | 4 | 995 | 5599381 | |
S72637.1 | - | 4 | 1048 | |||
X66872.1 | - | 4 | 1048 |