Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM184B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195071.1 | NP_001182000.1 | 4 | 638 | ||
NM_001195072.1 | NP_001182001.1 | 5 | 724 | |||
NM_012264.4 | NP_036396.2 | 4 | 623 | |||
XM_011530112.2 | XP_011528414.2 | 4 | 672 | |||
XM_011530113.1 | XP_011528415.1 | 4 | 602 | |||
XM_011530114.1 | XP_011528416.1 | 4 | 634 | |||
XM_011530115.2 | XP_011528417.1 | 3 | 393 | |||
XM_011530116.2 | XP_011528418.2 | 4 | 664 | |||
XM_011530117.1 | XP_011528419.1 | 5 | 700 | |||
XM_011530118.1 | XP_011528420.1 | 5 | 674 | |||
XM_011530119.1 | XP_011528421.1 | 3 | 428 | |||
XM_017028754.1 | XP_016884243.1 | 4 | 673 | |||
XM_017028755.1 | XP_016884244.1 | 4 | 666 | |||
XM_017028756.1 | XP_016884245.1 | 4 | 602 | |||
XM_017028757.1 | XP_016884246.1 | 3 | 390 | |||
XM_017028758.1 | XP_016884247.1 | 4 | 602 | |||
XM_017028759.1 | XP_016884248.1 | 3 | 428 | |||
GenBank mRNA | AB097053.1 | - | 4 | 594 | ||
AK075417.1 | - | 2 | 111 | |||
AK292652.1 | - | 4 | 623 | |||
AK309071.1 | - | 4 | 659 | |||
AL096879.1 | - | 3 | 316 | |||
BC015489.1 | - | 4 | 579 | 3896682 | ||
BI197128.1 | - | 3 | 317 | |||
BX648345.1 | - | 4 | 674 | |||
CR456413.1 | - | 3 | 457 | |||
CR749859.1 | - | 5 | 630 |