Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031613.1 | NP_113801.1 | 5, 6 | 674 | ||
XM_006243407.2 | XP_006243469.1 | 6, 7 | 1134 | |||
XM_006243408.2 | XP_006243470.1 | 6, 7 | 4336 | |||
XM_008766331.1 | XP_008764553.1 | 6, 7 | 726 | |||
GenBank mRNA | U59240.1 | - | 5, 6 | 674 |