Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130106.1 | NP_001123578.1 | 8, 9 | 1350 | ||
NM_001297600.1 | NP_001284529.1 | 6, 7 | 1265 | |||
NM_012267.4 | NP_036399.3 | 7, 8 | 1459 | |||
XM_005258700.2 | XP_005258757.1 | 7, 8 | 1168 | |||
XM_005258701.2 | XP_005258758.1 | 7, 8 | 1230 | |||
XM_011526691.1 | XP_011524993.1 | 8, 9 | 1326 | |||
XM_017026543.1 | XP_016882032.1 | 7, 8 | 1165 | |||
XM_017026544.1 | XP_016882033.1 | 7, 8 | 1187 | |||
XM_017026545.1 | XP_016882034.1 | 7, 8 | 1179 | |||
GenBank mRNA | AB020592.1 | - | 7, 8 | 1181 | ||
AF093420.1 | - | 7, 8 | 1184 | |||
AF187859.1 | - | 7, 8 | 1149 | |||
AF217996.1 | - | 8, 9 | 1336 | |||
AK075293.1 | - | 7, 8 | 1206 | |||
AK129755.1 | - | 7, 8 | 1262 | |||
AK130636.1 | - | 903 | ||||
AK130781.1 | - | 7, 8 | 1196 | |||
AK294358.1 | - | 6, 7 | 1264 | |||
BC001236.1 | - | 6, 7 | 1114 | 3454675 | ||
BC002373.2 | - | 7, 8 | 1171 | 2822836 | ||
CR457118.1 | - | 6, 7 | 1016 |