Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031622.2 | NP_113810.2 | 6 | 2646 | ||
XM_006243409.2 | XP_006243471.1 | 6 | 3140 | |||
GenBank mRNA | BC109380.1 | - | 6 | 2597 | 7446401 | |
M64301.1 | - | 5 | 2273 |