Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031629.1 | NP_113817.1 | 2 | 287 | ||
GenBank mRNA | FQ215475.1 | - | 2 | 305 | ||
FQ216787.1 | - | 2 | 304 | |||
FQ218364.1 | - | 2 | 306 | |||
FQ219781.1 | - | 2 | 306 | |||
FQ220578.1 | - | 2 | 304 | |||
FQ220909.1 | - | 2 | 306 | |||
FQ225189.1 | - | 2 | 344 | |||
FQ226073.1 | - | 2 | 305 | |||
FQ226711.1 | - | 2 | 305 | |||
FQ228014.1 | - | 2 | 305 | |||
FQ232850.1 | - | 2 | 306 | |||
L17127.1 | - | 2 | 287 |