Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099.4 | NP_001090.2 | 1 | 197 | ||
NM_001134194.1 | NP_001127666.1 | 1 | 197 | |||
NM_001292037.1 | NP_001278966.1 | 1 | 197 | |||
XM_011512946.1 | XP_011511248.1 | 1 | 161 | |||
XM_011512947.2 | XP_011511249.1 | 1 | 161 | |||
GenBank mRNA | AB102888.1 | - | 1 | 107 | ||
AK300540.1 | - | 1 | 158 | |||
BC007460.1 | - | 1 | 157 | 3951204 | ||
BC008493.1 | - | 1 | 154 | 4272289 | ||
BC016344.1 | - | 1 | 161 | 3950907 | ||
BC017877.1 | - | 1 | 147 | |||
DC383453.1 | - | 1 | 197 | |||
M24902.1 | - | 1 | 130 | |||
M34840.1 | - | 1 | 121 | |||
X52174.1 | - | 1 | 130 | |||
X53605.1 | - | 1 | 143 |