Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012275.2 | NP_036407.1 | 3, 4 | 274 | ||
NM_173170.1 | NP_775262.1 | 3, 4 | 246 | |||
GenBank mRNA | AF186094.1 | - | 3, 4 | 275 | ||
AF201830.1 | - | 2, 3 | 140 | |||
AF230377.1 | - | 2, 3 | 113 | |||
AJ242737.1 | - | 3, 4 | 194 | |||
AJ242738.1 | - | 3, 4 | 185 | |||
AK290249.1 | - | 3, 4 | 184 | |||
AY359117.1 | - | 3, 4 | 179 | |||
BC024747.1 | - | 3, 4 | 246 | 4996939 |