Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031647.2 | NP_113835.2 | 17, 18 | 2106 | ||
XM_006252656.2 | XP_006252718.1 | 17, 18 | 2392 | |||
XM_006252657.2 | XP_006252719.1 | 18, 19 | 2452 | |||
XM_006252658.1 | XP_006252720.1 | 17, 18 | 4364 | |||
XM_008771003.1 | XP_008769225.1 | 19, 20 | 3373 | |||
XM_008771004.1 | XP_008769226.1 | 18, 19 | 3313 | |||
XM_008771005.1 | XP_008769227.1 | 20, 21 | 2682 | |||
XM_008771006.1 | XP_008769228.1 | 19, 20 | 2546 | |||
GenBank mRNA | AB032164.1 | - | 17, 18 | 2100 | ||
BC081770.1 | - | 17, 18 | 2106 | 7132495 |