Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112712.1 | NP_001106183.1 | 13 | 1403 | ||
NM_001112713.1 | NP_001106184.1 | 13 | 1403 | |||
NM_031657.3 | NP_113845.2 | 13 | 1403 | |||
XM_006253653.2 | XP_006253715.1 | 13 | 1512 | |||
XM_008771504.1 | XP_008769726.1 | 12 | 1670 | |||
GenBank mRNA | AF040750.1 | - | 13 | 1403 | ||
X97439.1 | - | 13 | 1403 | |||
X97440.1 | - | 13 | 1403 | |||
Y09365.1 | - | 13 | 1431 |