Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032614.2 | NP_116003.2 | 8 | 811 | ||
XM_006230497.2 | XP_006230559.1 | 8 | 1224 | |||
GenBank mRNA | AF118651.1 | - | 8 | 645 | ||
BC086381.1 | - | 8 | 811 | 7301390 | ||
FQ216152.1 | - | 5 | 461 |