Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057099.3 | NP_476440.2 | 3 | 263 | ||
XM_008767811.1 | XP_008766033.1 | 3 | 294 | |||
GenBank mRNA | BC058451.1 | - | 3 | 267 | 6891542 | |
D10754.1 | - | 3 | 252 | |||
FQ209811.1 | - | 3 | 266 | |||
FQ217385.1 | - | 3 | 266 | |||
FQ224639.1 | - | 3 | 266 |
Gene Symbol: LOC100360846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729028.3 | XP_002729074.2 | 2 | 411 |