Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133381.3 | NP_596872.3 | 2 | 183 | ||
XM_008767524.1 | XP_008765746.1 | 2 | 313 | |||
GenBank mRNA | AB066219.1 | - | 2 | 160 | ||
AY462245.1 | - | 2 | 183 |