Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138878.2 | NP_620233.1 | 4 | 433 | ||
GenBank mRNA | AF095740.1 | - | 4 | 232 | ||
BC084728.1 | - | 4 | 253 | 7315541 | ||
CB581170.1 | - | 4 | 444 | |||
FQ230524.1 | - | 3 | 159 |