Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sele | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138879.1 | NP_620234.1 | 2 | 95 | ||
XM_008769608.1 | XP_008767830.1 | 3 | 477 | |||
GenBank mRNA | L25527.1 | - | 2 | 95 | ||
X69981.1 | - | 2 | 147 |